missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMG2L1 Antibody (OTI1H10), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71538CL1
This item is not returnable.
View return policy
Description
HMG2L1 Monoclonal antibody specifically detects HMG2L1 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| HMG2L1 | |
| Monoclonal | |
| CoraFluor 1 | |
| High mobility group protein 2-like 1, high-mobility group protein 2-like 1, HMG box domain containing 4, HMG domain-containing protein 4, HMG2L1HMG box-containing protein 4, HMGBCG, Protein HMGBCG, THC211630 | |
| Human recombinant protein fragment corresponding to amino acids 1-348 of human HMGXB4 (NP_005478) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI1H10 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 10042 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction