missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGA2 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98735H
This item is not returnable.
View return policy
Description
HMGA2 Polyclonal specifically detects HMGA2 in Human samples. It is validated for Western Blot.
Specifications
| HMGA2 | |
| Polyclonal | |
| Western Blot | |
| BABLHMGI-C, high mobility group AT-hook 2, High mobility group AT-hook protein 2, high mobility group protein HMGI-C, high-mobility group (nonhistone chromosomal) protein isoform I-C, High-mobility group protein HMGI-C, HMGICSTQTL9, LIPO | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human HMGA2. | |
| 0.1 mL | |
| Cancer | |
| 8091 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| HRP | |
| PBS | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction