missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB1/HMG-1 Antibody (19N12A1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-27400IR
This item is not returnable.
View return policy
Description
HMGB1/HMG-1 Monoclonal specifically detects HMGB1/HMG-1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| HMGB1/HMG-1 | |
| Monoclonal | |
| DyLight 755 | |
| P09429 | |
| HMGB1 | |
| Full-length recombinant human HMGB1 was used as the immunogen. | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Histones and Modified Histones, Neuroscience, Transcription Factors and Regulators | |
| 3146 | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 19N12A1 | |
| Western Blot | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| This antibody recognizes both B box domain (amino acids 88-162) and full length HMGB1 protein. | |
| Store at 4C in the dark. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction