missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGB1/HMG-1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2322CL1
This item is not returnable.
View return policy
Description
HMGB1/HMG-1 Polyclonal antibody specifically detects HMGB1/HMG-1 in Bovine,Canine,Human,Mouse,Rabbit,Rat,Sheep samples. It is validated for Flow Cytometry,ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Flow (Intracellular),Simple Western,KnockDown,Immunocytochemistry/Immunofluorescence
Specifications
| HMGB1/HMG-1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Bovine, Canine, Human, Mouse, Rabbit | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CoraFluor 1 | |
| Amphoterin, high mobility group box 1, High mobility group protein 1, high mobility group protein B1, high-mobility group (nonhistone chromosomal) protein 1, high-mobility group box 1, HMG-1, HMG1DKFZp686A04236, HMG3, SBP-1, Sulfoglucuronyl carbohydrate binding protein | |
| A synthetic peptide made to an internal portion of the human HMGB1 protein sequence (between residues 100-200). [UniProt #P09429] | |
| 0.1 mL | |
| Cellular Markers, Chromatin Research, Neuroscience | |
| 3146 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction