Learn More
Invitrogen™ HMGB4 Monoclonal Antibody (OTI5G3)
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA527031
Product Code. 15926624
  • € 570.00 / 100µL
Description
HMGB4 Monoclonal Antibody for Western Blot, IHC (P)
HMGB4 contains two HMG-box regions, which is found in a variety of eukaryotic chromosomal proteins and transcription.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HMGB4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8WW32
HMGB4
Full length human recombinant protein of HMGB4 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot
OTI5G3
Unconjugated
HMGB4
1700001F22Rik; AI326135; dJ1007G16.5; high mobility group box 4; high mobility group protein B4; high-mobility group box 4; HMG2 like; Hmgb4; LOC539195 protein
Mouse
Affinity Chromatography
RUO
127540
-20°C, Avoid Freeze/Thaw Cycles