missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA557370
This item is not returnable.
View return policy
Description
Immunogen sequence: PPSSFLLFCQ DHYAQLKREN PNWSVVQVAK ATGKMWSTAT DLEKHPYEQR VALLRAKYFE ELELYRKQCN ARKKYRMS Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 63%.
HMGB4 contains two HMG-box regions, which is found in a variety of eukaryotic chromosomal proteins and transcription.
Specifications
| HMGB4 | |
| Polyclonal | |
| Unconjugated | |
| HMGB4 | |
| 1700001F22Rik; AI326135; dJ1007G16.5; high mobility group box 4; high mobility group protein B4; high-mobility group box 4; HMG2 like; Hmgb4; LOC539195 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127540 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WW32 | |
| HMGB4 | |
| Recombinant protein corresponding to Human HMGB4. Recombinant protein control fragment (Product #RP-94470). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction