missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGB4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA579375
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: Rat Brain Tissue, Rat Testis Tissue, JURKAT Cell, HT1080 Cell.
HMGB4 contains two HMG-box regions, which is found in a variety of eukaryotic chromosomal proteins and transcription.
Specifications
| HMGB4 | |
| Polyclonal | |
| Unconjugated | |
| HMGB4 | |
| 1700001F22Rik; AI326135; dJ1007G16.5; high mobility group box 4; high mobility group protein B4; high-mobility group box 4; HMG2 like; Hmgb4; LOC539195 protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 127540, 685271, 69317 | |
| -20°C | |
| Lyophilized |
| Western Blot | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| Q6P8W9, Q8WW32 | |
| HMGB4 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human HMGB4 (24-41aa RNKFKEQQPNTYVGFKEF). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction