missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMGCL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-32767
This item is not returnable.
View return policy
Description
HMGCL Polyclonal specifically detects HMGCL in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| HMGCL | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| HMGCL | |
| Full length human HMGCL Recombinant protein. | |
| 100 μL | |
| Cancer, Signal Transduction | |
| 3155 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0mg/mL | |
| Western Blot 1:1000-1:10000, Immunohistochemistry 10 - 1:500, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 | |
| 3-hydroxy-3-methylglutaryl-CoA lyase, 3-hydroxymethyl-3-methylglutaryl-CoA lyase, EC 4.1.3.4, HLmitochondrial | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Tested in human. Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction