Learn More
HMGCL Antibody (S08-7H5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19654-50ul
 View more versions of this product
Product Code. 18685683
  • € 421.00 / 50µL
Description
HMGCL Monoclonal antibody specifically detects HMGCL in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HMGCL
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Western Blot
S08-7H5
Unconjugated
3-hydroxy-3-methylglutaryl-CoA lyase, 3-hydroxymethyl-3-methylglutaryl-CoA lyase, EC 4.1.3.4, HLmitochondrial
A synthetic peptide of human HMGCL (Uniprot # P35914)
50 μg
Cancer, Signal Transduction
3155
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.