missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HMGCS1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16696494
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16696494 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16696494 Supplier Invitrogen™ Supplier No. PA5100011

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total HMGCS1.

This enzyme condenses acetyl-CoA with acetoacetyl-CoA to form HMG-CoA, which is the substrate for HMG-CoA reductase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HMGCS1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene HMGCS1
Gene Accession No. P17425, Q01581, Q8JZK9
Gene Alias 3-hydroxy-3-methylglutaryl coenzyme A (HMG-CoA) synthase; 3-hydroxy-3-methylglutaryl coenzyme A synthase; 3-hydroxy-3-methylglutaryl-CoA synthase 1; 3-hydroxy-3-methylglutaryl-CoA synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble); 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble) isoform 1; 3-hydroxy-3-methylglutaryl-Coenzyme A synthase 1 (soluble) isoform 2; B130032C06Rik; HMG-CoA synthase; HMGCS; Hmgcs1; HMGCS1 protein-like; hydroxymethylglutaryl-CoA synthase 1; hydroxymethylglutaryl-CoA synthase, cytoplasmic; Unknown (protein for IMAGE:7955111)
Gene Symbols HMGCS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human HMGCS1(Accession Q01581), corresponding to amino acid residues K100-G150.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 208715, 29637, 3157
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.