missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HMGN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5106465
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HMG17.
The protein encoded by this gene binds nucleosomal DNA and is associated with transcriptionally active chromatin. Along with a similar protein, HMGN1, the encoded protein may help maintain an open chromatin configuration around transcribable genes. The protein has also been found to have antimicrobial activity against bacteria, viruses and fungi.
Specifications
| HMGN2 | |
| Polyclonal | |
| Unconjugated | |
| HMGN2 | |
| high mobility group nucleosomal binding domain 2; high mobility group nucleosome binding domain 2; high mobility group nucleosome-binding domain-containing protein 2; high mobility group protein 17; high mobility group protein N2; high-mobility group (nonhistone chromosomal) protein 17; high-mobility group nucleosomal binding domain 2; high-mobility group nucleosome-binding domain-containing protein 2; HMG17; Hmg-17; Hmgn2; Non-histone chromosomal protein HMG-17; nonhistone chromosomal protein HMG-17 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114637, 15331, 3151 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P05204, P09602, P18437 | |
| HMGN2 | |
| A synthesized peptide derived from human HMGN2(Accession P05204), corresponding to amino acid residues K5-G55. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction