Learn More
HMGXB4 Monoclonal Antibody (OTI1H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA801862S
Encompass_Preferred
 View more versions of this product
Product Code. 17520263
  • € 242.00 / 30µL
Description
HMGXB4 Monoclonal antibody specifically detects HMGXB4 in Human samples. It is validated for Western Blot
Specifications
Specifications
HMGXB4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
O00329
Mouse
Affinity chromatography
RUO
5293
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI1H10
Unconjugated
PIK3CD
high mobility group protein 2-like 1; high-mobility group protein 2-like 1; HMG box domain containing 4; HMG box-containing protein 4; HMG domain-containing protein 4; HMG2L1; HMGBCG; HMG-box containing 4; HMGXB4; Protein HMGBCG; Protein THC211630; THC211630
Human recombinant protein fragment corresponding to amino acids 286-610 of human PIK3CD produced in E.coli.