missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HMOX1 Monoclonal Antibody (GT664)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA531560

Product Code. 16374994

  • € 520.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Keep as concentrated solution. Predicted reactivity: Mouse (81%), Rat (80%), Dog (85%), Pig (82%), Bovine (83%). Positive Control: HeLa, HeLa 500 uM CoCl2 treatment for 24 hr. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HMOX1
Monoclonal
1 mg/mL
PBS with no preservative
P09601
HMOX1
Recombinant protein encompassing a sequence within the center region of human Heme Oxygenase 1. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG1
ELISA, Western Blot, Immunocytochemistry
GT664
Unconjugated
HMOX1
bK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 protein
Mouse
Protein G
RUO
3162
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.