Learn More
HMOX2 Mouse anti-Human, Clone: OTI1H10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503823
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Heme oxygenase, an essential enzyme in heme catabolism, cleaves heme to form biliverdin, which is subsequently converted to bilirubin by biliverdin reductase, and carbon monoxide, a putative neurotransmitter. Heme oxygenase activity is induced by its substrate heme and by various nonheme substances. Heme oxygenase occurs as 2 isozymes, an inducible heme oxygenase-1 and a constitutive heme oxygenase-2. HMOX1 and HMOX2 belong to the heme oxygenase family. Alternative splice variants encoding the same protein have been identified at this locus.Specifications
| HMOX2 | |
| Monoclonal | |
| Unconjugated | |
| HMOX2 | |
| HO-2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3163 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P30519 | |
| HMOX2 | |
| Full length human recombit protein of human HMOX2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.