missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HMOX2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14817-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an epithelial-derived, integral membrane serine protease. This protease forms a complex with the Kunitz-type serine protease inhibitor, HAI-1, and is found to be activated by sphingosine 1-phosphate. This protease has been shown to cleave and activate hepatocyte growth factor/scattering factor, and urokinase plasminogen activator, which suggest the function of this protease as an epithelial membrane activator for other proteases and latent growth factors. The expression of this protease has been associated with breast, colon, prostate, and ovarian tumors, which implicates its role in cancer invasion, and metastasis.Specifications
| HMOX2 | |
| Polyclonal | |
| Unconjugated | |
| HMOX2 | |
| heme oxygenase (decycling) 2, Heme oxygenase 2, HMOX2, HO 2, HO2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 15369, 3163, 79239 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70252, P23711, P30519 | |
| HMOX2 | |
| HMOX2 Fusion Protein Ag6575 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction