missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HN1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521779
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, A431, HeLa, K562, THP-1, HL-60. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
HN1 (hematopoietic- and neurologic-expressed sequence 1) is a highly conserved protein that is expressed in developing and regenerating tissues. Overexpression of HN1 has been reported in various tumors like glioma and breast cancer. Hn1 has also been reported as a marker for human ovarian carcinoma and can distinguish epithelial ovarian carcinoma cells from normal ovarian surface epithelial cells. This collective information suggests that HN1 is involved in processes associated with cell proliferation, repair and/or growth.
Specifications
| HN1 | |
| Polyclonal | |
| Unconjugated | |
| JPT1 | |
| Androgen-regulated protein 2; ARM2; hematological and neurological expressed 1; hematological and neurological expressed 1 protein; hematological and neurological expressed sequence 1; HN1; HN1A; Jpt1; Jupiter microtubule associated homolog 1; Jupiter microtubule associated homolog 1, N-terminally processed | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51155 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 1.22 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q9UK76 | |
| JPT1 | |
| Recombinant protein encompassing a sequence within the center region of human HN1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction