missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HN1L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30492
This item is not returnable.
View return policy
Description
HN1L Polyclonal antibody specifically detects HN1L in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| HN1L | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 90861 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| C16orf34, chromosome 16 open reading frame 34, CRAMP_1 like, FLJ13092, hematological and neurological expressed 1-like, hematological and neurological expressed 1-like protein, HN1 like, HN1-like protein, KIAA1426, L11 | |
| Between 50 and 100 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction