missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-3 alpha/FoxA1 Antibody (rFOXA1/1515), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08920MFV450
This item is not returnable.
View return policy
Description
HNF-3 alpha/FoxA1 Monoclonal antibody specifically detects HNF-3 alpha/FoxA1 in Human, Rat samples. It is validated for Flow Cytometry
Specifications
| HNF-3 alpha/FoxA1 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Flow Cytometry | |
| rFOXA1/1515 | |
| Flow Cytometry | |
| forkhead box A1, Forkhead box protein A1, hepatocyte nuclear factor 3, alpha, hepatocyte nuclear factor 3-alpha, HNF-3A, HNF-3-alpha, HNF3ATCF-3A, MGC33105, TCF3A, Transcription factor 3A | |
| Recombinant human HNF-3 alpha/FoxA1 protein fragment (around aa372-472) (exact sequence is proprietary) (Uniprot: P55317) | |
| 0.1 mL | |
| Breast Cancer, Cancer, Stem Cell Markers | |
| 3169 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction