missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-4 alpha/NR2A1 Antibody (843716), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB4605C
This item is not returnable.
View return policy
Description
HNF-4 alpha/NR2A1 Monoclonal antibody specifically detects HNF-4 alpha/NR2A1 in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Specifications
| HNF-4 alpha/NR2A1 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry (Intracellular Staining), Immunocytochemistry | |
| 843716 | |
| Western Blot, Simple Western, Intracellular Staining by Flow Cytometry, Immunocytochemistry | |
| FLJ39654, hepatic nuclear factor 4 alpha, hepatocyte nuclear factor 4, alpha, hepatocyte nuclear factor 4-alpha, HNF4a7, HNF-4-alpha, HNF4alpha10/11/12, HNF4HNF4a8, MODY, MODY1, NR2A1HNF4a9, NR2A21, Nuclear receptor subfamily 2 group A member 1, TCF, TCF-14, TCF14HNF4alpha, Transcription factor 14, Transcription factor HNF-4, transcription factor-14 | |
| E. coli-derived recombinant human HNF-4 alpha/NR2A1, Val130-Ser330, Accession # P41235 | |
| 0.1 mL | |
| Stem Cell Markers | |
| 3172 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction