missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF-6/ONECUT1 Antibody (OTI4F12), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70906CL1
This item is not returnable.
View return policy
Description
HNF-6/ONECUT1 Monoclonal antibody specifically detects HNF-6/ONECUT1 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| HNF-6/ONECUT1 | |
| Monoclonal | |
| CoraFluor 1 | |
| HNF6Ahepatocyte nuclear factor 6, alpha, HNF6hepatocyte nuclear factor 6, HNF-6One cut domain family member 1, one cut homeobox 1one cut domain, family member 1 | |
| Human recombinant protein fragment corresponding to amino acids 85-198 of human HNF6 (NP_004489) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunocytochemistry/Immunofluorescence | |
| OTI4F12 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 3175 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction