missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98696-100ul
This item is not returnable.
View return policy
Description
HNF1 Polyclonal antibody specifically detects HNF1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| HNF1 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| HNF1 homeobox A, IDDM20, LFB1transcription factor 1, hepatic; LF-B1, hepatic nuclear factor (HNF1), albuminproximal factor, MODY3, TCF1hepatic, Transcription factor 1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HNF1. | |
| 100 μg | |
| Transcription Factors and Regulators | |
| 6927 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction