missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF4A Monoclonal Antibody (OTI8B8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806576
This item is not returnable.
View return policy
Description
Hepatocyte nuclear factor 4 alpha (HNF4 alpha) is an orphan nuclear receptor. HNF4a is expressed in the liver, kidney, intestine, and pancreas. Mutation of HNF4a in humans has been associated with maturity-onset diabetes of the young type 1 (MODY1). HNF4 binds to DNA as an exclusive homodimer. The HNF4a gene is alternatively spliced and may generate up to nine different isoforms, HNF4a1 through HNF4a9.Specifications
| HNF4A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P41235 | |
| HNF4A | |
| Human recombit protein fragment corresponding to amino acids 1-315 of human HNF4A produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI8B8 | |
| Unconjugated | |
| HNF4A | |
| FRTS4, HNF4, HNF4a7, HNF4a8, HNF4a9, HNF4alpha, MODY, MODY1, NR2A1, NR2A21, TCF, TCF14 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 3172 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction