missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNF4A Monoclonal Antibody (OTI9H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA806583S
This item is not returnable.
View return policy
Description
HNF4A Monoclonal antibody specifically detects HNF4A in Human samples. It is validated for Western BlotSpecifications
| HNF4A | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P41235 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3172 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI9H8 | |
| Unconjugated | |
| HNF4A | |
| alpha transcription factor 4; FLJ39654; FRTS4; hepatic nuclear factor 4 alpha; hepatic nuclear factor 4, alpha; hepatocyte nuclear factor 4 alpha; hepatocyte nuclear factor 4, alpha; hepatocyte nuclear factor 4-alpha; HNF4; Hnf-4; HNF4 alpha; Hnf4a; HNF4a7; HNF4a8; HNF4a9; Hnf4alpha; HNF-4-alpha; HNF4-alpha; HNF4alpha10; HNF4alpha10/11/12; HNF4alpha11; HNF4alpha12; MODY; MODY1; NR2A1; NR2A21; Nuclear receptor 2A1; nuclear receptor subfamily 2 group A member 1; TCF; TCF14; TCF-14; Transcription factor 14; transcription factor HNF-4 | |
| Human recombinant protein fragment corresponding to amino acids 1-315 of human HNF4A produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction