missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hnRNP A1L2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5145164
This item is not returnable.
View return policy
Description
Immunogen sequence: GNFGGRSSGP YGGGGQYFAK PQNQGG Antigen sequence identity: Mouse 96%; Rat 96%.
Involved in the packaging of pre-mRNA into hnRNP particles, transport of poly(A) mRNA from the nucleus to the cytoplasm and may modulate splice site selection. [UniProt].
Specifications
| hnRNP A1L2 | |
| Polyclonal | |
| Unconjugated | |
| HNRNPA1L2 | |
| heterogeneous nuclear ribonucleoprotein A1-like 2; hnRNP A1-like 2; hnRNP core protein A1-like 2; HNRNPA1L; HNRNPA1L2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 144983 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q32P51 | |
| HNRNPA1L2 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 279-304. Recombinant protein control fragment (Product #RP-109934). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction