missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP C1 + C2 Antibody (9G1), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 355.00 - € 568.00
Specifications
| Antigen | hnRNP C1 + C2 |
|---|---|
| Clone | 9G1 |
| Dilution | Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30032321
|
Novus Biologicals
NBP3-26221-50ul |
50 μL |
€ 355.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30032080
|
Novus Biologicals
NBP3-26221-100ul |
100 μL |
€ 568.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
hnRNP C1 + C2 Monoclonal antibody specifically detects hnRNP C1 + C2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, ImmunocytochemistrySpecifications
| hnRNP C1 + C2 | |
| Western Blot 1:500-1:5000, Flow Cytometry, ELISA, Immunohistochemistry 1:50-1:200, Immunoprecipitation 1:200-1:1000, Immunocytochemistry/Immunofluorescence 1:20-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C1, C2, heterogeneous nuclear ribonucleoprotein C (C1/C2), heterogeneous nuclear ribonucleoproteins C1/C2, HNRNP, hnRNP C1/C2, hnRNPC, HNRPC, MGC104306, MGC105117, MGC117353, MGC131677, nuclear ribonucleoprotein particle C1 protein, nuclear ribonucleoprotein particle C2 protein, SNRPC | |
| A synthesized peptide derived from Human hnRNP C1 + C2 [UniProt P07910] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 9G1 | |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 3183 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title