Learn More
hnRNP C1 + C2 Antibody (CL2596) [mFluor Violet 450 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36777MFV450
Product Code. 30298420
  • € 759.00 / 0.10mL
Description
hnRNP C1 + C2 Monoclonal antibody specifically detects hnRNP C1 + C2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
hnRNP C1 + C2
Monoclonal
mFluor Violet 450 SE
C1, C2, heterogeneous nuclear ribonucleoprotein C (C1/C2), heterogeneous nuclear ribonucleoproteins C1/C2, HNRNP, hnRNP C1/C2, hnRNPC, HNRPC, MGC104306, MGC105117, MGC117353, MGC131677, nuclear ribonucleoprotein particle C1 protein, nuclear ribonucleoprotein particle C2 protein, SNRPC
This antibody was developed using a recombinant protein derived from P07910, with the exact immunogen sequence remaining proprietary.
0.1 mL
DNA Repair, DNA replication Transcription Translation and Splicing
3183
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL2596
50mM Sodium Borate
Mouse
Protein A purified