missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hnRNP CL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116648
This item is not returnable.
View return policy
Description
Positive Samples: THP-1, MCF7 Immunogen sequence: NLEKIEKEQS KQEVEVKNAK SEEEQSSSSM KKDETHVKME SEGGAEDSAE EGDPLDDDVN EDQGDDQLEL IKDDEKEAEE GEDDRDSTNG QDDS.
May play a role in nucleosome assembly by neutralizing basic proteins such as A and B core hnRNPs.
Specifications
| hnRNP CL1 | |
| Polyclonal | |
| Unconjugated | |
| HNRNPCL1 | |
| heterogeneous nuclear ribonucleoprotein C-like 1; hnRNP C-like-1; hnRNP core protein C-like 1; HNRNPCL1; HNRPCL1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 343069 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O60812 | |
| HNRNPCL1 | |
| A synthetic peptide corresponding to a sequence within amino acids 200 to the C-terminus of human HNRNPCL1 (NP_0010136531). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction