missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ hnRNP H3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17911601
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17911601 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17911601 Supplier Invitrogen™ Supplier No. PA5144017

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Jurkat whole cell, human PC-3 whole cell, human Raji whole cell, human K562 whole cell, human Caco-2 whole cell, human MCF-7 whole cell, human HL-60 whole cell, human PC-3 whole cell. IHC: human breast cancer tissue, human gastric cancer lymph node tissue, human liver cancer tissue, human ovarian serous adenocarcinoma tissue, human pancreatic cancer tissue, human placenta tissue, human rectal cancer tissue. ICC/IF: MCF-7 cell. Flow: K562 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

HNRPH3 belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein has two repeats of quasi-RRM domains that bind to RNAs. It is localized in nuclear bodies of the nucleus. This protein is involved in the splicing process and it also participates in early heat shock-induced splicing arrest by transiently leaving the hnRNP complexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP H3
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and no preservative
Gene HNRNPH3
Gene Accession No. P31942
Gene Alias 2H9; AA693301; AI666703; heterogeneous nuclear ribonucleoprotein 2H9; Heterogeneous nuclear ribonucleoprotein H3; heterogeneous nuclear ribonucleoprotein H3 (2H9); hnRNP 2H9; hnRNP H3; HNRNPH3; HNRPH3; Unknown (protein for MGC:136953); zgc:136953
Gene Symbols HNRNPH3
Host Species Rabbit
Immunogen E.coli-derived human HNRNPH3 recombinant protein (Position: K29-N268).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3189
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.