missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ hnRNP LL Polyclonal Antibody
GREENER_CHOICE

Product Code. 16536221
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16536221 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16536221 Supplier Invitrogen™ Supplier No. PA587841

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: IRNDNDSWDY TKPYLGRRDR GKGRQRQAIL GEHPSSFRHD GYGSHGPLLP LPSRYRMGSR DTPELVAYPL PQASSSYMHG GNPSGSVVMV SGLHQLKMNC S; Positive Samples: A-549, DU145, HepG2, 293T, Mouse liver.

HNRNPLL is a master regulator of activation-induced alternative splicing in T cells. In particular, it alters splicing of CD45 (PTPRC), a tyrosine phosphatase essential for T-cell development and activation (Oberdoerffer et al., 2008).
TRUSTED_SUSTAINABILITY

Specifications

Antigen hnRNP LL
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.68 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Hnrnpll
Gene Accession No. Q8WVV9, Q921F4
Gene Alias 2510028H02Rik; 2810036L13Rik; AI256697; AI852082; BLOCK24; heterogeneous nuclear ribonucleoprotein L like; heterogeneous nuclear ribonucleoprotein L-like; Hnrnpll; HNRPLL; RGD1305861; SRRF; stromal RNA regulating factor; stromal RNA-regulating factor
Gene Symbols Hnrnpll
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 250-350 of human HNRPLL (NP_6124032).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 72692, 92906
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.