missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP M Antibody (OTI4C9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71200F
This item is not returnable.
View return policy
Description
hnRNP M Monoclonal specifically detects hnRNP M in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| hnRNP M | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| HNRNPM | |
| Recombinant protein fragment corresponding to amino acids 1-261 of human hnRNP M (NP_112480) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Immunohistochemistry | |
| OTI4C9 | |
| Immunohistochemistry | |
| DKFZp547H118, heterogeneous nuclear ribonucleoprotein M, hnRNP M, HNRNPM4, HNRPM4heterogenous nuclear ribonucleoprotein M, HNRPMheterogenous nuclear ribonucleoprotein M4, HTGR1, M4 protein, N-acetylglucosamine receptor 1, NAGR1hnRNA-binding protein M4 | |
| Mouse | |
| Affinity Purified | |
| Cancer | |
| 4670.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction