missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hnRNP R Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100559
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total HNRNPR.
This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs and also contains a nuclear localization motif. Multiple alternatively spliced transcript variants have been found for this gene.
Specifications
| hnRNP R | |
| Polyclonal | |
| Unconjugated | |
| HNRNPR | |
| 2610003J05Rik; 2610528B01Rik; FLJ25714; heterogeneous nuclear ribonucleoprotein R; hnRNP R; HNRNPR; hnRNP-R; HNRPR | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10236, 319110, 74326 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O43390 | |
| HNRNPR | |
| A synthesized peptide derived from human HNRNPR(Accession O43390), corresponding to amino acid residues R585-K633. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction