missing translation for 'onlineSavingsMsg'
Learn More
Learn More
hnRNP U Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2135C
This item is not returnable.
View return policy
Description
hnRNP U Polyclonal antibody specifically detects hnRNP U in Human, Mouse samples. It is validated for Western Blot
Specifications
| hnRNP U | |
| Polyclonal | |
| Western Blot | |
| heterogeneous nuclear ribonucleoprotein U, heterogeneous nuclear ribonucleoprotein U (scaffold attachment factor A), hnRNP U, HNRPU, p120, p120 nuclear protein, pp120, SAFA, SAF-AhnRNPU, Scaffold attachment factor A, U21.1 | |
| The immunogen recognized by this antibody maps to a region between residue 775 and the C-terminus (residue 824) of human Heterogeneous Nuclear Ribonucleoprotein U using the numbering given in entry NP_114032.1 (GeneID 3192). | |
| 0.1 mL | |
| Cellular Markers | |
| 3192 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction