Learn More
hnRNP U Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2135PE
This item is not returnable.
View return policy
Description
hnRNP U Polyclonal antibody specifically detects hnRNP U in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation