missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ hnRNP UL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584305
This item is not returnable.
View return policy
Description
Immunogen sequence: QDSEKSKPAG SDGERRGVKR QRDEKDEHGR AYYEFREEAY HSRSKSPLPP EEEAKDEEED QTLVNLDTYT SDLHFQVSKD RYGGQPLFSE KFPTLWSGAR Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.
HNRNPUL2 gene ontology annotations related to this gene include RNA binding.
Specifications
| hnRNP UL2 | |
| Polyclonal | |
| Unconjugated | |
| HNRNPUL2 | |
| 1110031M08Rik; AI465155; heterogeneous nuclear ribonucleoprotein U like 2; heterogeneous nuclear ribonucleoprotein U-like 2; heterogeneous nuclear ribonucleoprotein U-like protein 2; Hnrnpul2; Hnrpul2; Manp; MLF1-associated nuclear protein; SAF-A2; Scaffold-attachment factor A2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 221092 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunofluorescence | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q1KMD3 | |
| HNRNPUL2 | |
| Recombinant protein corresponding to Human HNRNPUL2. Recombinant protein control fragment (Product #RP-106721). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction