missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPA0 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22293DL594
This item is not returnable.
View return policy
Description
HNRNPA0 Polyclonal antibody specifically detects HNRNPA0 in Human, Mouse samples. It is validated for Western Blot
Specifications
| HNRNPA0 | |
| Polyclonal | |
| Western Blot | |
| heterogeneous nuclear ribonucleoprotein A0, hnRNA binding protein, hnRNPA0, HNRPA0hnRNP A0 | |
| The immunogen this antibody was made to, maps to a region between residue 150 to 200 of human Heterogeneous Nuclear Ribonucleoprotein A0 using the numbering given in entry NP_006796.1 (GeneID 10949). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 10949 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction