missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPA0 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38221-100ul
This item is not returnable.
View return policy
Description
HNRNPA0 Polyclonal antibody specifically detects HNRNPA0 in Human,Mouse,Rat samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| HNRNPA0 | |
| Polyclonal | |
| Western Blot 1:500 - 1:1000, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| heterogeneous nuclear ribonucleoprotein A0, hnRNA binding protein, hnRNPA0, HNRPA0hnRNP A0 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human HNRNPA0 (NP_006796.1).,, Sequence:, MENSQLCKLFIGGLNVQTSESGLRGHFEAFGTLTDCVVVVNPQTKRSRCFGFVTYSNVEEADAAMAASPHAVDGNTVELKRAVSREDSARPGAHAKVKKLFVGGLKGDVAEGDLIEHFSQFGTVEKAEIIADKQSGKKRGFGFVYFQNHDAADKAAVVKFHPIQGHRVEVKKAVPKEDIY | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 10949 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction