missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPA0 Monoclonal Antibody (OTI8H8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809308S
This item is not returnable.
View return policy
Description
HNRNPA0 Monoclonal antibody specifically detects HNRNPA0 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| HNRNPA0 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96RI1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 9971 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI8H8 | |
| Unconjugated | |
| NR1H4 | |
| 1110055B05Rik; 3010025E17Rik; Heterogeneous nuclear ribonucleoprotein A0; hnRNA binding protein; hnRNP A0; HNRNPA0; Hnrpa0 | |
| Human recombinant protein fragment corresponding to amino acids 21-297 of human NR1H4 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction