missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HNRNPA0 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16539331
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16539331 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16539331 Supplier Invitrogen™ Supplier No. PA596492

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MENSQLCKLF IGGLNVQTSE SGLRGHFEAF GTLTDCVVVV NPQTKRSRCF GFVTYSNVEE ADAAMAASPH AVDGNTVELK RAVSREDSAR PGAHAKVKKL FVGGLKGDVA EGDLIEHFSQ FGTVEKAEII ADKQSGKKRG FGFVYFQNHD AADKAAVVKF HPIQGHRVEV KKAVPKEDIY; Positive Samples: MCF-7, BT-474, SKOV3, 293T, HeLa, Mouse brain, Mouse spleen, Rat brain; Cellular Location: Nucleus.

HNRPA0 belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind RNAs, followed by a glycine-rich C-terminus. This gene belongs to the A/B subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has two repeats of quasi-RRM domains that bind RNAs, followed by a glycine-rich C-terminus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HNRNPA0
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 5.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Hnrnpa0
Gene Accession No. Q13151, Q9CX86
Gene Alias 1110055B05Rik; 3010025E17Rik; Heterogeneous nuclear ribonucleoprotein A0; hnRNA binding protein; hnRNP A0; HNRNPA0; Hnrpa0
Gene Symbols Hnrnpa0
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human HNRNPA0 (NP_0067961).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10949, 498696, 77134
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.