missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12770-1-AP-150UL
This item is not returnable.
View return policy
Description
hnRNP D/AUF1 binds with high affinity to RNA molecules that contain AU-rich elements (AREs) found within the 3′ untranslated regions of many protooncogenes and cytokine mRNAs. It also binds to double and single stranded DNA sequences in a specific manner and functions a transcription factor. Each of the RNA-binding domains specifically can bind solely to a single stranded nonmonotonous 5′-UUAG-3′ sequence and also weaker to the single-stranded 5′-TTAGGG-3′ telomeric DNA repeat. It may be involved in translationally coupled mRNA turnover (referenced from swissprot).Specifications
| HNRNPD | |
| Polyclonal | |
| Unconjugated | |
| HNRNPD | |
| AUF1, AUF1A, hnRNP D0, HNRNPD, hnRNPD0, HNRPD, P37 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11991, 3184, 79256 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q14103, Q60668, Q9JJ54 | |
| HNRNPD | |
| HNRNPD Fusion Protein Ag3458 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction