missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPH1 Monoclonal Antibody (OTI2D11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810866S
This item is not returnable.
View return policy
Description
HNRNPH1 Monoclonal antibody specifically detects HNRNPH1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| HNRNPH1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UQ07, Q9WVS4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 26448, 5891 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2D11 | |
| Unconjugated | |
| MOK | |
| AI642080; E430005G16Rik; Heterogeneous nuclear ribonucleoprotein H; Heterogeneous nuclear ribonucleoprotein H, N-terminally processed; heterogeneous nuclear ribonucleoprotein H1; heterogeneous nuclear ribonucleoprotein H1 (H); hnRNP H; Hnrnph; HNRNPH1; HNRPH; HNRPH1; Ratsg1 | |
| Human recombinant protein fragment corresponding to amino acids 205-419 of human RAGE produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction