missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HNRNPH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14774-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the GTP cyclohydrolase family. The encoded protein is the first and rate-limiting enzyme in tetrahydrobiopterin biosynthesis, catalyzing the conversion of GTP into 7,8-dihydroneopterin triphosphate. BH4 is an essential cofactor required by aromatic amino acid hydroxylases as well as nitric oxide synthases. Mutations in this gene are associated with maligt hyperphenylalaninemia and dopa-responsive dystonia. Several alternatively spliced transcript variants encoding different isoforms have been described; however, not all variants give rise to a functional enzyme.Specifications
| HNRNPH1 | |
| Polyclonal | |
| Unconjugated | |
| HNRNPH1 | |
| hnRNP H, hnRNPH, HNRNPH1, HNRPH, HNRPH1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 140931, 3187, 59013 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O35737, P31943, Q8VHV7 | |
| HNRNPH1 | |
| HNRNPH1 Fusion Protein Ag6506 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction