missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HO-1/HMOX1/HSP32 Antibody (6B8-2F2), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59338FR
This item is not returnable.
View return policy
Description
HO-1/HMOX1/HSP32 Monoclonal specifically detects HO-1/HMOX1/HSP32 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| HO-1/HMOX1/HSP32 | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HMOX1 | |
| His-tagged Rat HO-1 | |
| 100 μL | |
| Primary | |
| Detects 32kDa. Does not cross-react with HO-2. | |
| Store at 4C in the dark. | |
| IgG1 κ |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 6B8-2F2 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
| Mouse | |
| Protein G purified | |
| Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism | |
| 3162 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction