missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HO-1/HMOX1/HSP32 Antibody (CL11661), Novus Biologicals™
Mouse Monoclonal Antibody
€ 329.00 - € 516.00
Specifications
| Antigen | HO-1/HMOX1/HSP32 |
|---|---|
| Clone | CL11661 |
| Dilution | Immunohistochemistry 1:500-1:1000, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671955
|
Novus Biologicals
NBP3-21199-100ul |
100 μg |
€ 516.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18658214
|
Novus Biologicals
NBP3-21199-25ul |
25 μg |
€ 329.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HO-1/HMOX1/HSP32 Monoclonal antibody specifically detects HO-1/HMOX1/HSP32 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| HO-1/HMOX1/HSP32 | |
| Immunohistochemistry 1:500-1:1000, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bK286B10, EC 1.14.99.3, heat shock protein, 32-kD, heme oxygenase (decycling) 1, HO, HO1, HO-1heme oxygenase 1, HSP32 | |
| This antibody was developed against a Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence of P09601. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL11661 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cellular Markers, ER Markers, Lipid and Metabolism | |
| PBS, pH 7.2, 40% glycerol | |
| 3162 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title