Learn More
Invitrogen™ HO-1 Monoclonal Antibody (6B8-2F2), PerCP
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA545348
Product Code. 17857072
  • € 455.00 / 100µg
Description
1 μg/mL of MA5-45348 was sufficient for detection of HO-1 in 10 μg of rat kidney lysate by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Detects approximately 32kDa. Does not cross-react with HO-2.
Heme oxygenase (HO) is a microsomal enzyme that catalyzes the oxidation of heme to the antioxidant molecules, biliverdin and carbon monoxide. HO consists of two homologous isozymes, an inducible HO-1 and a constitutively expressed HO-2. HO-1 is induced by a wide variety of stimuli including conditions of oxidative stress, inflammatory agents, transforming growth factor beta and heat shock. The increase in expression of HO-1 is thought to be a cellular defense mechanism against oxidative stress since elevated HO could eventually generate more bilirubin, an anti-oxidant.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HO-1
Monoclonal
1 mg/mL
2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative
P06762, P09601, P14901
HMOX1
His-tagged Rat HO-1.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG1 κ
Western Blot, Immunocytochemistry
6B8-2F2
PerCP
HMOX1
bK286B10; D8Wsu38e; heat shock protein 32; heat shock protein, 32-kD; heme orygenase 1; heme oxygenase; heme oxygenase (decycling) 1; heme oxygenase (decyclizing) 1; heme oxygenase 1; heme oxygenase-1; Hemox; hemoxygenase; Heox; HEOXG; Hmox; Hmox1; HMOX1D; HO; Ho1; HO-1; Hsp32; P32 protein
Mouse
Protein G
RUO
15368, 24451, 3162
4°C