missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HO-2/HMOX2 Antibody (OTI1C2), DyLight 650, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70895C
This item is not returnable.
View return policy
Description
HO-2/HMOX2 Monoclonal specifically detects HO-2/HMOX2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| HO-2/HMOX2 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HMOX2 | |
| Full length recombinant protein of human HO-2/HMOX2 (NP_002125) produced in HEK293T cells. | |
| 0.1 mL | |
| 3163.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI1C2 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| heme oxygenase (decycling) 2, heme oxygenase 2, HO2, HO-2EC 1.14.99.3 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction