Learn More
MedChemExpress Hoechst 33342 trihydrochloride
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light ∣ The compound is unstable in solutions, freshly prepared is recommended. |
| Quantity | 50 mg |
| Shipping Condition | Room temperature |
| Solubility | DMSO : ≥ 46 mg/mL (81.86 mM) ∣ H2O : ≥ 5.6 mg/mL (9.97 mM) |
| Molecular Weight (g/mol) | 561.93 g/mol |
| CAS | 875756-97-1 |
| Synonym | bisBenzimide H 33342 trihydrochloride ∣ HOE 33342 trihydrochloride |