Learn More
Hoefer™ Cams for SE 400 and SE 600 Series Units and SE 6015 Gel Caster
Click to view available options
For Use With (Equipment):
SE 400, SE 600 Series Vertical Electrophoresis Unit, SE 6015 Gel Caster
Unit Size:
Pack of 4
Description
- For use with SE 400 and SE 600 Series Units and SE 6015 Gel Caster
Specifications
Specifications
| Color | Black |
| For Use With (Equipment) | SE 400, SE 600 Series Vertical Electrophoresis Unit, SE 6015 Gel Caster |
| Type | Cams |