Learn More
Hoefer™ Gel Casters for Mini Vertical Electrophoresis Systems
Easy-to-use casters
Brand: Hoefer™ SE235
Additional Details : Weight : 1,468.00000kg
Description
SE245 Dual-Gel Caster
- For use with SE250, SE260, or MiniVE Vertical Electrophoresis Systems
- Suitable for single concentration and gradient gels
- SE235 4-Gel Caster is for use with SE260 or MiniVE Electrophoresis Systems
- SE 275 4-Gel Caster and SE215 10-Gel Caster are for use with SE250 Electrophoresis System
Specifications
SE 260, MiniVE™ Electrophoresis System | |
Gel Caster | |
Caster body with gasket, face plate with inlet port, 10 x 10.5cm rectangular Glass plates, 10 x 10.5cm notched Alumina plates, spring clamps, Wax Paper, space-saver plate, Polycarbonate filler sheets, black Vinyl cap, filler plug set | |
1 to 4 |