Learn More
Hoefer™ SUB25 Maxi-Plus Submarine Gel Electrophoresis Combs
For use with standard and cooled gel electrophoresis models
Brand: Hoefer™ SUB25-C2-26MC
Additional Details : Weight : 0.01000kg
Description
- Comb sample volumes listed are for 5 mm deep wells
Specifications
Standard and Cooled Gel Electrophoresis Model | |
8 | |
0.2 cm | |
0.27 in. | |
0.19 in. |
Comb | |
0.07 in. | |
60 μL | |
0.7 cm | |
0.5 cm |