missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HOGA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06562-100ul
This item is not returnable.
View return policy
Description
HOGA1 Polyclonal antibody specifically detects HOGA1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HOGA1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| 4-hydroxy-2-oxoglutarate aldolase 1, C10orf65, chromosome 10 open reading frame 65, DHDPS2, DHDPSLHP3, DHDPS-like protein, Dihydrodipicolinate synthase-like, dihydrodipicolinate synthase-like, mitochondrial, dihydrodipicolinate synthetase homolog 2, EC 4.1.3.16, FLJ37472, FLJ56960, N-acetylneuraminate pyruvate lyase 2 (putative), NPL2, Probable 2-keto-4-hydroxyglutarate aldolase, probable 4-hydroxy-2-oxoglutarate aldolase, mitochondrial, Probable KHG-aldolase, Protein 569272 | |
| Produced in rabbits immunized with E. coli-derived Human HOGA1 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 112817 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction